Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
TC0400003900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_001109684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_001109684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_001109685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_001109685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_144531.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032461.mm.1 |
NM_144531.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
TC0400003900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_001109684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_001109684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_001109685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_001109685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_144531.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032462.mm.1 |
NM_144531.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
TC0400003900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_001109684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_001109684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_001109685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_001109685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_144531.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032460.mm.1 |
NM_144531.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032467.mm.1 |
TC0400003900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032467.mm.1 |
NM_001109685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032467.mm.1 |
NM_001109685.1 |
Refseq |