Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033165.3 | NP_001028337.2 | 2 - 3 | 521 | 80 | |
GenBank mRNA | AK008359.1 | - | 3 - 4 | 275 | 80 | |
AK008425.1 | - | 3 - 4 | 551 | 80 | ||
AK136359.1 | - | 2 - 3 | 215 | 80 | ||
BC119039.1 | - | 2 - 3 | 286 | 8733788 | 80 | |
BC120693.1 | - | 2 - 3 | 286 | 40129616 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |