Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029401.1 | NP_083677.1 | 3 - 4 | 397 | 56 | |
GenBank mRNA | AK016425.1 | - | 3 - 4 | 418 | 56 | |
BC057376.1 | - | 2 - 3 | 269 | 56 | ||
BC132477.1 | - | 3 - 4 | 397 | 40130754 | 56 | |
BC137851.1 | - | 3 - 4 | 397 | 8860870 | 56 | |
BC144974.1 | - | 4 - 5 | 452 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |