Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028878.3 | NP_083154.1 | 3 - 4 | 541 | 69 | |
GenBank mRNA | AB096860.1 | - | 3 - 4 | 626 | 69 | |
AJ633679.1 | - | 3 - 4 | 550 | 69 | ||
AK014544.1 | - | 3 - 4 | 542 | 69 | ||
AK028784.1 | - | 3 - 4 | 542 | 69 | ||
AK128923.1 | - | 3 - 4 | 541 | 69 | ||
AK162399.1 | - | 2 - 3 | 286 | 69 | ||
BC107411.1 | - | 3 - 4 | 542 | 40059042 | 69 | |
BC107412.2 | - | 3 - 4 | 546 | 40059050 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |