Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163288.2 | NP_001156760.2 | 11 - 12 | 1894 | 77 | |
GenBank mRNA | AK041075.1 | - | 2 - 3 | 172 | 77 | |
BC125408.1 | - | 1 - 2 | 52 | 77 | ||
BC125410.1 | - | 1 - 2 | 52 | 77 | ||
BC157977.1 | - | 11 - 12 | 1819 | 9088138 | 77 | |
BC158124.1 | - | 11 - 12 | 1871 | 9007393 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |