Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025900.2 | NP_080176.2 | 2 - 3 | 316 | 137 | |
GenBank mRNA | AK012615.1 | - | 2 - 3 | 257 | 137 | |
AK012877.1 | - | 2 - 3 | 251 | 137 | ||
AK033451.1 | - | 2 - 3 | 206 | 137 | ||
AK154013.1 | - | 2 - 3 | 298 | 137 | ||
BC048844.1 | - | 2 - 3 | 266 | 137 | ||
BC055451.1 | - | 2 - 3 | 262 | 4164972 | 137 | |
BU920290.1 | - | 2 - 3 | 300 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |