Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026382.2 | NP_080658.2 | 5 - 6 | 618 | 113 | |
GenBank mRNA | AK007431.1 | - | 5 - 6 | 607 | 113 | |
AK012916.1 | - | 5 - 6 | 613 | 113 | ||
AK018315.1 | - | 5 - 6 | 613 | 113 | ||
AK167282.1 | - | 1 - 2 | 1208 | 113 | ||
BC037073.1 | - | 5 - 6 | 608 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |