Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167939.1 | NP_001161411.1 | 5 - 6 | 622 | 80 | |
NM_028993.4 | NP_083269.4 | 5 - 6 | 622 | 80 | ||
GenBank mRNA | AK018662.1 | - | 5 - 6 | 590 | 80 | |
AK034837.1 | - | 5 - 6 | 584 | 80 | ||
AK039196.1 | - | 5 - 6 | 588 | 80 | ||
AK049277.1 | - | 5 - 6 | 583 | 80 | ||
AK081967.1 | - | 5 - 6 | 623 | 80 | ||
AK164005.1 | - | 5 - 6 | 589 | 80 | ||
BC125568.1 | - | 5 - 6 | 622 | 40130083 | 80 | |
BC132139.1 | - | 5 - 6 | 622 | 40130416 | 80 | |
BC144990.1 | - | 5 - 6 | 622 | 9053538 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |