Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024922.2 | NP_001020093.2 | 2 - 3 | 254 | 63 | |
GenBank mRNA | AK139073.1 | - | 2 - 3 | 266 | 63 | |
BC085301.1 | - | 2 - 3 | 254 | 63 | ||
BC099554.1 | - | 2 - 3 | 261 | 30308124 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |