Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024922.2 | NP_001020093.2 | 3 - 4 | 338 | 79 | |
GenBank mRNA | AK139073.1 | - | 3 - 4 | 350 | 79 | |
AK179358.1 | - | 1 - 2 | 53 | 79 | ||
AK214595.1 | - | 1 - 2 | 63 | 79 | ||
BC085301.1 | - | 3 - 4 | 338 | 79 | ||
BC099554.1 | - | 3 - 4 | 345 | 30308124 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |