Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024922.2 | NP_001020093.2 | 8 - 9 | 948 | 129 | |
GenBank mRNA | BC085301.1 | - | 8 - 9 | 948 | 129 | |
BC099554.1 | - | 8 - 9 | 955 | 30308124 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |