Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027182.2 | NP_081458.1 | 1 - 2 | 280 | 78 | |
GenBank mRNA | AK010336.1 | - | 1 - 2 | 242 | 78 | |
AK142463.1 | - | 253 | 78 | |||
AK146877.1 | - | 1 - 2 | 281 | 78 | ||
AK151568.1 | - | 1 - 2 | 216 | 78 | ||
BC064315.1 | - | 1 - 2 | 203 | 78 | ||
BC126946.1 | - | 1 - 2 | 179 | 40090292 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |