Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009951.1 | NP_001009951.1 | 8 - 9 | 1020 | 102 | |
GenBank mRNA | AK220415.1 | - | 7 - 8 | 976 | 102 | |
BC088983.1 | - | 8 - 9 | 1020 | 5700628 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |