Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008278.2 | NP_032304.2 | 1 - 2 | 159 | 85 | |
GenBank mRNA | AK146038.1 | - | 1 - 2 | 127 | 85 | |
BB870447.1 | - | 1 - 2 | 160 | 85 | ||
BC021157.1 | - | 1 - 2 | 118 | 4208980 | 85 | |
U44389.1 | - | 1 - 2 | 118 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |