Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166397.1 | NP_001159869.1 | 3 - 4 | 673 | 75 | |
NM_001166398.1 | NP_001159870.1 | 3 - 4 | 616 | 75 | ||
GenBank mRNA | AK014329.1 | - | 3 - 4 | 449 | 75 | |
AK034185.1 | - | 3 - 4 | 449 | 75 | ||
AK034621.1 | - | 3 - 4 | 447 | 75 | ||
AK043440.1 | - | 3 - 4 | 449 | 75 | ||
AK044926.1 | - | 3 - 4 | 447 | 75 | ||
AK081299.1 | - | 3 - 4 | 392 | 75 | ||
AK172960.1 | - | 3 - 4 | 673 | 75 | ||
BC049088.1 | - | 3 - 4 | 428 | 75 | ||
BC052383.1 | - | 3 - 4 | 422 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |