Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Invs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281977.1 | NP_001268906.1 | 15 - 16 | 3195 | 70 | |
NM_001281978.1 | NP_001268907.1 | 17 - 18 | 3473 | 70 | ||
NM_010569.4 | NP_034699.3 | 15 - 16 | 3247 | 70 | ||
GenBank mRNA | AF034860.3 | - | 15 - 16 | 3189 | 70 | |
AJ010902.1 | - | 15 - 16 | 3188 | 70 | ||
AK041671.1 | - | 13 - 14 | 2907 | 70 | ||
AK144794.1 | - | 3 - 4 | 1177 | 70 | ||
AK158766.1 | - | 17 - 18 | 3432 | 70 | ||
BC126929.1 | - | 15 - 16 | 3084 | 40081431 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |