Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029572.2 | NP_083848.1 | 1 - 2 | 204 | 154 | |
GenBank mRNA | AK003217.1 | - | 1 - 2 | 173 | 154 | |
AK151497.1 | - | 1 - 2 | 205 | 154 | ||
AK160113.1 | - | 1 - 2 | 187 | 154 | ||
AK172973.1 | - | 146 | 154 | |||
BC019558.1 | - | 1 - 2 | 111 | 4192664 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |