Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012044.2 | NP_036174.1 | 3 - 4 | 342 | 107 | |
GenBank mRNA | AF112984.1 | - | 3 - 4 | 342 | 107 | |
AF166098.1 | - | 3 - 4 | 338 | 107 | ||
AK148187.1 | - | 358 | 107 | |||
BC027524.1 | - | 2 - 3 | 231 | 1533423 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |