Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025637.3 | NP_079913.2 | 4 - 5 | 806 | 123 | |
NM_028456.3 | NP_082732.2 | 2 - 3 | 590 | 123 | ||
GenBank mRNA | AF439556.1 | - | 2 - 3 | 589 | 123 | |
AF439557.1 | - | 3 - 4 | 796 | 123 | ||
AK014130.1 | - | 2 - 3 | 528 | 123 | ||
AK134288.1 | - | 2 - 3 | 581 | 123 | ||
BC128287.1 | - | 2 - 3 | 589 | 40102432 | 123 | |
BC128288.1 | - | 4 - 5 | 805 | 123 | ||
BC139340.1 | - | 2 - 3 | 589 | 8862362 | 123 | |
BC139342.1 | - | 2 - 3 | 589 | 8862364 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |