Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 2 - 3 | 543 | 82 | |
NM_001290652.1 | NP_001277581.1 | 2 - 3 | 543 | 82 | ||
GenBank mRNA | AK031490.1 | - | 2 - 3 | 522 | 82 | |
AK040521.1 | - | 2 - 3 | 545 | 82 | ||
BC056382.1 | - | 2 - 3 | 233 | 6853936 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |