Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 13 - 14 | 2089 | 77 | |
NM_019867.2 | NP_063920.2 | 11 - 12 | 1726 | 77 | ||
GenBank mRNA | AJ238898.1 | - | 11 - 12 | 1707 | 77 | |
AK138433.1 | - | 11 - 12 | 1695 | 77 | ||
AY038025.1 | - | 11 - 12 | 1701 | 77 | ||
BC022680.1 | - | 5 - 6 | 504 | 77 | ||
BC039279.1 | - | 13 - 14 | 2089 | 4973826 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |