Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174853.2 | NP_777278.2 | 8 - 9 | 1814 | 81 | |
GenBank mRNA | AF513724.1 | - | 8 - 9 | 1811 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |