Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076062.1 | - | 4 - 5 | 583 | 64 | |
AK146766.1 | - | 4 - 5 | 591 | 64 | ||
AK158633.1 | - | 4 - 5 | 580 | 64 | ||
AK162294.1 | - | 4 - 5 | 544 | 64 | ||
AK168629.1 | - | 4 - 5 | 517 | 64 | ||
AK169317.1 | - | 4 - 5 | 596 | 64 | ||
BC054722.1 | - | 4 - 5 | 525 | 5694816 | 64 | |
BC138301.1 | - | 4 - 5 | 579 | 8861320 | 64 | |
BC138302.1 | - | 4 - 5 | 580 | 8861321 | 64 | |
D29016.1 | - | 4 - 5 | 533 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |