Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008112.4 | NP_032138.3 | 5 - 6 | 814 | 95 | |
GenBank mRNA | AK007871.1 | - | 5 - 6 | 757 | 95 | |
AK078144.1 | - | 757 | 95 | |||
AK078514.1 | - | 5 - 6 | 761 | 95 | ||
AK079690.1 | - | 4 - 5 | 655 | 95 | ||
AK083063.1 | - | 5 - 6 | 756 | 95 | ||
AK090083.1 | - | 5 - 6 | 762 | 95 | ||
AK137761.1 | - | 5 - 6 | 761 | 95 | ||
AK143633.1 | - | 5 - 6 | 759 | 95 | ||
AK143681.1 | - | 5 - 6 | 793 | 95 | ||
AK149958.1 | - | 5 - 6 | 767 | 95 | ||
AK150356.1 | - | 5 - 6 | 749 | 95 | ||
AK150655.1 | - | 5 - 6 | 784 | 95 | ||
AK152250.1 | - | 5 - 6 | 748 | 95 | ||
AK153381.1 | - | 5 - 6 | 727 | 95 | ||
AK159734.1 | - | 5 - 6 | 759 | 95 | ||
AK166317.1 | - | 5 - 6 | 759 | 95 | ||
AK167659.1 | - | 5 - 6 | 771 | 95 | ||
AK169299.1 | - | 5 - 6 | 725 | 95 | ||
BC024971.1 | - | 5 - 6 | 735 | 95 | ||
BC053381.1 | - | 2914 | 95 | |||
BC055341.1 | - | 5 - 6 | 740 | 6808475 | 95 | |
L36314.1 | - | 5 - 6 | 620 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |