Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2006008 |
AK160167.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2006008 |
AK160456.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2006008 |
BC011140.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2006008 |
BC104333.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2006008 |
AK003930.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562240 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
762864 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
249562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
566266 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
377981 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
538041 |
BC048651.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
538041 |
NM_207258.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359473 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858140 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515461 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
550897 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219162 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838683 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
525537 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250207 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930730 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
287933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
565227 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828312 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
947784 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546750 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
15879 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66810 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
89842 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113756 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105866 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2006008 |
AK160167.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2006008 |
AK160456.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2006008 |
BC011140.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2006008 |
BC104333.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2006008 |
AK003930.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P181297 |
AK004500.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931708 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931712 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931731 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931705 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931701 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931706 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28931720 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830661 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830722 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830712 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830702 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830684 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07830677 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105866 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P181297 |
AK004500.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133978 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105866 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07830702 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11011491 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10714376 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11011494 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11011511 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10714371 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11011498 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11011481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28931627 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931700 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931681 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931690 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931667 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11099041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931673 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931786 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10967822 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931775 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931783 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11011511 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931605 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11085390 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10798602 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931729 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10967827 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11099040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931611 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931785 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931606 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931701 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11085398 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931776 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10714368 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931749 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931684 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931708 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931682 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10469796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931600 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10714370 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931671 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931731 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931697 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11011512 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11075985 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931740 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931788 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931748 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931695 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931719 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10720171 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931607 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931710 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931668 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931677 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931595 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931703 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931713 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10798629 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931686 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931752 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931675 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28931746 |
- |
- |