Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439974 |
AK050107.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439974 |
AK050212.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439974 |
BC025515.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439974 |
BC030735.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439974 |
NM_146020.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
92882 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393561 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393550 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393553 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393557 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393551 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24393569 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05875924 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01312753 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05726701 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05813403 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05874071 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05874068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24393554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24393550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24393551 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393568 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393551 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393560 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393553 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393549 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393557 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05813403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393554 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393550 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24393561 |
- |
- |