Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007748.3 | NP_031774.1 | 1 - 2 | 118 | 172 | |
GenBank mRNA | AK002402.1 | - | 1 - 2 | 120 | 172 | |
AK019318.1 | - | 1 - 2 | 123 | 172 | ||
AK166496.1 | - | 1 - 2 | 120 | 172 | ||
BC003807.1 | - | 1 - 2 | 118 | 3487598 | 172 | |
BC052816.1 | - | 1 - 2 | 124 | 6439987 | 172 | |
L06465.1 | - | 1 - 2 | 117 | 172 | ||
U08440.1 | - | 1 - 2 | 107 | 172 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |