Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016835.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016830.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016830.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016830.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016828.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016828.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016828.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016829.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016829.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016829.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016834.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016834.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016834.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016832.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016832.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016832.mm.1 |
TC1500002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016831.mm.1 |
NM_001033275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016831.mm.1 |
NM_001033275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016831.mm.1 |
TC1500002092.mm.1 |
MTA TC |