Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013756.2 | NP_038784.1 | 1 - 2 | 73 | 122 | |
GenBank mRNA | AF092929.1 | - | 1 - 2 | 70 | 122 | |
BC114344.2 | - | 1 - 2 | 70 | 40049236 | 122 | |
BC116176.2 | - | 1 - 2 | 74 | 40049224 | 122 | |
BC116177.1 | - | 1 - 2 | 73 | 40049232 | 122 | |
BC134358.1 | - | 1 - 2 | 74 | 40049234 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |