Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700021054.mm.1 |
TC0700002374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021054.mm.1 |
NM_001101586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021054.mm.1 |
NM_001101586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021051.mm.1 |
TC0700002374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021051.mm.1 |
NM_001101586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021051.mm.1 |
NM_001101586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021050.mm.1 |
TC0700002374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021050.mm.1 |
NM_001101586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021050.mm.1 |
NM_001101586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021052.mm.1 |
TC0700002374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021052.mm.1 |
NM_001101586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021052.mm.1 |
NM_001101586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021053.mm.1 |
TC0700002374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021053.mm.1 |
NM_001101586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021053.mm.1 |
NM_001101586.2 |
Refseq |