Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500023757.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023757.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023757.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023759.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023759.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023759.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023764.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023764.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023764.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023766.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023766.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023766.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023761.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023761.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023761.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023767.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023767.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023767.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023765.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023765.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023765.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023758.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023758.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023758.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023760.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023760.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023760.mm.1 |
NM_001029867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023762.mm.1 |
TC0500002692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023762.mm.1 |
NM_001029867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023762.mm.1 |
NM_001029867.1 |
Refseq |