Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010584.3 | NP_034714.1 | 5 - 6 | 637 | 129 | |
GenBank mRNA | AK007447.1 | - | 5 - 6 | 637 | 129 | |
AY157364.1 | - | 5 - 6 | 621 | 129 | ||
AY619693.1 | - | 5 - 6 | 620 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |