Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10425041 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430181 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430185 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430182 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10425042 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10425044 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10425045 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430184 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10425043 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430183 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10430180 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10430179 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10425040 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1500004605.mm.1 |
NM_001134802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004605.mm.1 |
NM_001134802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004605.mm.1 |
TC1500000631.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1436271_at |
- |
- |
Mouse Expression Set 430 |
1436271_PM_at |
- |
- |