Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880581 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880579 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880583 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880578 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880582 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880585 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880580 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
207693 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
565300 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
962466 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AF100424.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AF100425.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AF189772.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AK019491.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AK028481.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AK035151.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AK083939.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
AK144174.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
BC004032.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
BC004746.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
BC029196.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
BC037116.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
BC060097.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735885 |
NM_013758.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
937039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648206 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545000 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
643661 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542829 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633474 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
125548 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P112210 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880579 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880583 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880586 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880557 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880571 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880569 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880580 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880554 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880578 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880582 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880568 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880564 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880561 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880576 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880574 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880589 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880565 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880590 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880572 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880559 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880556 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880555 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880600 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29880601 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112210 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08228425 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08228420 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P549348 |
NM_198601.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P549348 |
AK029521.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P100473 |
NM_198601.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P100473 |
AK029521.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123260 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08228420 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112210 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542509 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542494 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542504 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542515 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542487 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11542485 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880583 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880585 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880582 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880579 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880580 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880578 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29880581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29880583 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880534 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880605 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880606 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11542490 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880590 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11542491 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880596 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880613 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880564 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880586 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880614 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880572 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880537 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880562 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11402283 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11455747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11255439 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11455745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880557 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880617 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880615 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880532 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880531 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880552 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880603 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11519457 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880545 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880535 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11263205 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880600 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880559 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880553 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880561 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880542 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880579 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880592 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880528 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880527 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11263207 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880576 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880556 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880593 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880598 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11519454 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880555 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11575657 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880581 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880571 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880580 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880530 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880566 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880568 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880595 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880574 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880549 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11714028 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880546 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880569 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29880608 |
- |
- |