Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020752.1 | - | 549 | 88 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |