Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290414.1 | NP_001277343.1 | 1 - 2 | 188 | 76 | |
GenBank mRNA | AF199608.1 | - | 1 - 2 | 188 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |