Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160307.1 | NP_001153779.1 | 2 - 3 | 159 | 135 | |
GenBank mRNA | AK052580.1 | - | 2 - 3 | 160 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |