Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA016947.1 | - | 1 | 144 | 77 | |
AA475384.1 | - | 1 | 152 | 77 | ||
AA980874.1 | - | 4 | 356 | 77 | ||
AK140913.1 | - | 3 | 1598 | 77 | ||
BC019774.1 | - | 4 | 579 | 77 | ||
BC031815.1 | - | 1 | 184 | 4166503 | 77 | |
BC055868.1 | - | 1 | 144 | 4190595 | 77 | |
BE625312.1 | - | 1 | 149 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |