Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700013373.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013373.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013373.mm.1 |
NM_011975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013371.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013371.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013371.mm.1 |
NM_011975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013361.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013361.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013361.mm.1 |
NM_011975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013370.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013370.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013370.mm.1 |
NM_011975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013376.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013376.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013376.mm.1 |
NM_011975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013364.mm.1 |
TC0700001612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013364.mm.1 |
NM_011975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013364.mm.1 |
NM_011975.3 |
Refseq |