Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101516.1 | NP_001094986.1 | 1 | 1021 | 150 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK041859.1 | - | Chr. - - [-] on Build | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |