Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019643.3 | NP_062617.2 | 5 - 6 | 669 | 104 | |
GenBank mRNA | AK046519.1 | - | 5 - 6 | 576 | 104 | |
AK145384.1 | - | 5 - 6 | 680 | 104 | ||
AK166569.1 | - | 5 - 6 | 675 | 104 | ||
BC076599.1 | - | 5 - 6 | 669 | 30614283 | 104 | |
U64033.1 | - | 5 - 6 | 758 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |