Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ren1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031192.3 | NP_112469.1 | 3 - 4 | 400 | 140 | |
GenBank mRNA | AK052685.1 | - | 3 - 4 | 400 | 140 | |
AK085309.1 | - | 3 - 4 | 400 | 140 | ||
BC061053.1 | - | 3 - 4 | 398 | 30313659 | 140 | |
X16642.1 | - | 3 - 4 | 382 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |