Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178786.4 | NP_848901.2 | 6 - 7 | 1117 | 150 | |
GenBank mRNA | AK028550.1 | - | 6 - 7 | 1169 | 150 | |
AK028795.1 | - | 6 - 7 | 1118 | 150 | ||
AK035748.1 | - | 6 - 7 | 1035 | 150 | ||
AK162813.1 | - | 7 - 8 | 1132 | 150 | ||
EF494897.1 | - | 6 - 7 | 916 | 150 | ||
EF494898.1 | - | 6 - 7 | 916 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |