Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700012591.mm.1 |
NM_001142539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012591.mm.1 |
NM_001142539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012591.mm.1 |
TC1700001400.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013104.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013104.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013104.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013105.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013105.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013105.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013102.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013102.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013102.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012590.mm.1 |
NM_001142539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012590.mm.1 |
NM_001142539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012590.mm.1 |
TC1700001400.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013109.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013109.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013109.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012589.mm.1 |
NM_001142539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012589.mm.1 |
NM_001142539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012589.mm.1 |
TC1700001400.mm.1 |
MTA TC |