Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010831.1 | NP_001010831.1 | 1 | 635 | 113 | |
GenBank mRNA | BC139084.1 | - | 1 | 684 | 8862106 | 113 |
BC139085.1 | - | 1 | 683 | 8862107 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |