Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb2b26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY293281.1 | - | 2 | 249 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Amplicon spans exons and probe does not span exons |