Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_001013360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_001013360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_001013362.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_001013362.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_030689.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
NM_030689.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014846.mm.1 |
TC1500001887.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500014858.mm.1 |
NM_001013362.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014858.mm.1 |
NM_001013362.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014858.mm.1 |
NM_028763.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014858.mm.1 |
NM_028763.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014858.mm.1 |
TC1500001887.mm.1 |
MTA TC |