Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900005153.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005153.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005153.mm.1 |
TC0900000699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005158.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005158.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005158.mm.1 |
TC0900000699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005154.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005154.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005154.mm.1 |
TC0900000699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005157.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005157.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005157.mm.1 |
TC0900000699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005155.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005155.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005155.mm.1 |
TC0900000699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005156.mm.1 |
NM_001014398.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005156.mm.1 |
NM_001014398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005156.mm.1 |
TC0900000699.mm.1 |
MTA TC |