Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014398.2 | NP_001014420.2 | 9 - 10 | 2278 | 86 | |
GenBank mRNA | AY701870.1 | - | 9 - 10 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |