Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017966.2 | NP_001017966.1 | 6 - 7 | 1113 | 75 | |
GenBank mRNA | AK129015.1 | - | 1091 | 75 | ||
BC021415.1 | - | 6 - 7 | 1094 | 75 | ||
BF468816.1 | - | 1 - 2 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |